Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, PVDF, Straight, Sanitary Clamp to Threaded Adapter, 1"" Ladish Clamp x 1/2"" NPT(M)
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX3180859
Encompass
Tri-Clamp male thread adapters allow quick and easily installation of your sanitary set-up to an NPT male pipe thread. Though both materials may be autoclaved, PVDF tolerates high temperatures better than PP does and will perform more consistently after repeated autoclaving. Clamps and gaskets required for a complete system are sold separately.
Catalog No. NC1471497