Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,2-Trichloro-1,2,2-trifluoroethane, Reagent, 99.8%, Spectrum™ Chemical
SDP

Catalog No. 18600027 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
120 mL
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-600-027 120 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-600-027 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. TR113120ML

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

TR113, 76-13-1, C2Cl3F3

Spectrum™ Chemical 1,1,2-Trichloro-1,2,2-trifluoroethane, Reagent , is a chlorofluorocarbon that is very unreactive, and if released will stay in the atmosphere for long periods of time. Also referred to as CFC-113, it was one of the many CFCs that were used as refrigerants, aerosol propellants and cleansing agent for electrical and electronic components. It has today mostly been replaced in laboratory analytics by other solvents but is none the less still used in small doses for some of its unique properties. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.

Chemical Identifiers

CAS 76-13-1
Molecular Formula C2Cl3F3
Molecular Weight (g/mol) 187.37
InChI Key AJDIZQLSFPQPEY-UHFFFAOYSA-N
IUPAC Name 1,1,2-trichloro-1,2,2-trifluoroethane
SMILES FC(F)(Cl)C(F)(Cl)Cl

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 120 mL
Percent Purity 99.8%
Grade Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.