Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,2-Trichloro-3,3,3-trifluoro-1-propene, 97%, Thermo Scientific™

Catalog No. AAL0926303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0926303 1 g
AAL0926306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0926303 Supplier Thermo Scientific Chemicals Supplier No. L0926303

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 431-52-7
Molecular Formula C3Cl3F3
Molecular Weight (g/mol) 199.378
MDL Number MFCD00013672
InChI Key QSSVZVNYQIGOJR-UHFFFAOYSA-N
Synonym 1,1,2-trichloro-3,3,3-trifluoropropene, 1,1,2-trichloro-3,3,3-trifluoro-1-propene, 1-propene, 1,1,2-trichloro-3,3,3-trifluoro, propene, 1,1,2-trichloro-3,3,3-trifluoro, trifluorotrichloropropene, acmc-20ap8g, 1,2-trichlorotrifluoro-1-propene, 1,1,2-trichlorotrifluoro-1-propene, 1,1-trifluoro-2,3,3-trichloropropene, 1,2-trichloro-3,3,3-trifluoropropene
PubChem CID 67938
IUPAC Name 1,1,2-trichloro-3,3,3-trifluoroprop-1-ene
SMILES C(=C(Cl)Cl)(C(F)(F)F)Cl

Specifications

Melting Point -114°C
Density 1.617
Boiling Point 88°C
Refractive Index 1.406
Quantity 1 g
UN Number UN2810
Beilstein 1766481
Formula Weight 199.39
Percent Purity 97%
Chemical Name or Material 1,1,2-Trichloro-3,3,3-trifluoro-1-propene
missing translation for 'healthHazard1' H302+H312-H331
missing translation for 'healthHazard2' GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
missing translation for 'healthHazard3' P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P311-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 207-075-0
RTECSNumber UD2696000
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.