Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,1,2-Trichloroethane

Catalog No. 502903937
Encompass
Change view
Click to view available options
Quantity:
25 mL
250 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-290-3937 1 L
50-185-9817 250 mL
50-185-9818 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-290-3937 Supplier Sigma Aldrich Supplier No. 4662121L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1,1,2-trichloroethane (1,1,2-tca), a chlorinated aliphatic hydrocarbon, is a common groundwater pollutant
  • Microbial detoxification of 1,1,2-tca has been studied
  • Ni supported on mesoporous silica (sba-16) catalyzed hydrodechlorination of 1,1,2-trichloroethane has been reported
  • Rotational isomers of 1,1,2-trichloroethane in the gaseous and liquid states have been investigated by the infrared absorption spectra
TRUSTED_SUSTAINABILITY

Specifications

CAS 79-00-5
Density 1.435 g/mL (at 25°C (literature))
Boiling Point 110°C to 115°C (lit.)
Molecular Formula C2H3Cl3
Refractive Index n20/D 1.471 (literature)
Linear Formula ClCH2CHCl2
MDL Number MFCD00000852
Quantity 1 L
Molecular Weight (g/mol) 133.4
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.