Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,1,3,3-Tetramethyl-1,3-Diphenyldisilazane

Catalog No. 36515710G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-515-710G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 36-515-710G Supplier Sigma Aldrich Supplier No. 36515710G
Only null left
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1,1,3,3-Tetramethyl-1,3-diphenyldisilazane can be used:As a starting material for the synthesis of its substitution products with the tetrachlorides of silicon and tin.To prepare Cu-, Zn- and H2-phthalocyanines using phthalimide.As a pre-column deactivation agent prior to the determination of the fatty acidsby GC-MS.

Specifications

CAS 174484-84-5
Density 0.954 g/mL at 25°C
Boiling Point 67°C
Molecular Formula C12H22ClMgNSi2
Refractive Index n20/D 1.538
Linear Formula [(CH3)3Si]2NC6H4MgCl
MDL Number MFCD00134370
Quantity 10 g
Synonym 1,3-Diphenyl-1,1,3,3-tetramethyldisilazane; DPTMDS
Molecular Weight (g/mol) 296.24 g/mol
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.