Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,1,3,3-Tetramethylbutyl isocyanide

Catalog No. 2264915G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-649-15G 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-649-15G Supplier Sigma Aldrich Supplier No. 2264915G
May include imposed supplier surcharges.
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1,1,3,3-Tetramethylbutyl isocyanide

Specifications

CAS 14542-93-9
Density 0.794 g/mL (at 25°C (literature))
Boiling Point 55°C to 57°C (11 mmHg)
Molecular Formula C9H17N
Refractive Index n20/D 1.422 (literature)
Linear Formula (CH3)3 CCH2C(CH3)2 NC
MDL Number MFCD00000003
Quantity 250 mg
Synonym tert-Octyl isocyanide; Walborskys reagent
Molecular Weight (g/mol) 139.24
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.