Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,1,3,3-Tetramethylguanidine

Catalog No. 2417685ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-176-85ML 5 mL
24-176-8100ML 100 mL
24-176-8500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 24-176-85ML Supplier Sigma Aldrich Supplier No. 2417685ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Promotes the pentavalent bismuth oxidation of primary and secondary alchohols to aldehydes and ketones.

Specifications

CAS 80-70-6
Density 0.918 g/mL at 25°C
Boiling Point 52°C to 54°C (11 mmHg, lit.)
Molecular Formula C5H13N3
Refractive Index n20/D 1.469
Linear Formula (CH3)2NC(=NH)N(CH3)2
MDL Number MFCD00008323
Quantity 5 mL
Synonym N,N,N',N'-Tetramethylguanidine
Molecular Weight (g/mol) 115.18 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.