Learn More
Medchemexpress LLC 1-(1,3-benzothiazol-2-yl)-1-(2,4-dimethylphenyl)ethanol | 1253901-26-6 | MFCD20527324 | 100.0% | 283.39 g/mol | C17H17NOS | 10 MG
SDP
Catalog No. 5000228373
Encompass_Preferred
AC-265347 is a small-molecule calcium-sensing receptor (CaSR) agonist and positive allosteric modulator (ago-PAM) used for biochemical and cellular research to probe CaSR signaling and related pharmacology.
  • CaSR agonist and positive allosteric modulator for receptor pharmacology studies.
  • High purity suitable for analytical and cellular assays (reported 99.97%).
  • Molecular formula C17H17NOS and molecular weight 283.39 g/mol.
  • Available in multiple milligram pack sizes for in vitro experiments; commonly used at micromolar concentrations.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-28373 Supplier Medchemexpress LLC Supplier No. HY11785110MG
AC-265347 is a small-molecule calcium-sensing receptor (CaSR) agonist and positive allosteric modulator (ago-PAM) used for biochemical and cellular research to probe CaSR signaling and related pharmacology.
  • CaSR agonist and positive allosteric modulator for receptor pharmacology studies.
  • High purity suitable for analytical and cellular assays (reported 99.97%).
  • Molecular formula C17H17NOS and molecular weight 283.39 g/mol.
  • Available in multiple milligram pack sizes for in vitro experiments; commonly used at micromolar concentrations.
  • Intended for research use only; not for human or clinical use.