Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-(1,3-benzothiazol-2-yl)-1-(2,4-dimethylphenyl)ethanol | 1253901-26-6 | MFCD20527324 | 99.97% | 283.39 g/mol | C17H17NOS | 25 MG
SDP

Catalog No. 5000228019
Encompass_Preferred

AC-265347 is a calcium-sensing receptor (CaSR) agonist and positive allosteric modulator used in preclinical research on hyperparathyroidism and CaSR pharmacology. It is supplied as a high-purity small molecule suitable for in vitro and in vivo studies.

  • High purity (99.97%).
  • Molecular weight 283.39 g/mol; formula C17H17NOS.
  • Soluble in DMSO at 100 mg/mL for in vitro applications.
  • Documented in vivo formulations produce clear solutions at ≥2.5 mg/mL.
  • Powder stable at -20°C for up to 3 years under recommended storage.

Catalog No. 50-002-28019 Supplier Medchemexpress LLC Supplier No. HY11785125MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AC-265347 is a calcium-sensing receptor (CaSR) agonist and positive allosteric modulator used in preclinical research on hyperparathyroidism and CaSR pharmacology. It is supplied as a high-purity small molecule suitable for in vitro and in vivo studies.

  • High purity (99.97%).
  • Molecular weight 283.39 g/mol; formula C17H17NOS.
  • Soluble in DMSO at 100 mg/mL for in vitro applications.
  • Documented in vivo formulations produce clear solutions at ≥2.5 mg/mL.
  • Powder stable at -20°C for up to 3 years under recommended storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.