Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(1,3-Benzothiazol-2-yl)-2-bromo-1-ethanone, 90+%, Thermo Scientific™

Catalog No. CC06005CB
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC06005CB 250 mg
CC06005DA 1 g
CC06005EA 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC06005CB Supplier Thermo Scientific Chemicals Supplier No. CC06005CB
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 54223-20-0
Molecular Formula C9H6BrNOS
Molecular Weight (g/mol) 256.12
MDL Number MFCD03659698
InChI Key AYWGYNKWZWBMSV-UHFFFAOYSA-N
Synonym 1-1,3-benzothiazol-2-yl-2-bromo-1-ethanone, 1-benzo d thiazol-2-yl-2-bromoethanone, 1-1,3-benzothiazol-2-yl-2-bromoethanone, 2-bromoacetyl benzothiazole, 1-benzothiazol-2-yl-2-bromoethanone, 1-benzothiazol-2-yl-2-bromo-ethanone, 1-1,3-benzothiazol-2-yl-2-bromoethan-1-one, pubchem23327, 2-bromoacetyl-1,3-benzothiazole
PubChem CID 2776256
SMILES BrCC(=O)C1=NC2=CC=CC=C2S1

Specifications

Assay Percent Range 90%
Quantity 250 mg
IUPAC Name 1-(1,3-benzothiazol-2-yl)-2-bromoethanone
Formula Weight 256.12
Percent Purity 90%
Chemical Name or Material 1-(1, 3-Benzothiazol-2-yl)-2-bromo-1-ethanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.