Learn More
Medchemexpress LLC 1-(1-(3-methoxyphenyl)-7-propoxyindolizin-3-yl)ethan-1-one | 1693766-04-9 | 99.5% | 323.39 g·mol⁻¹ | C20H21NO3 | 25 MG
SDP
Catalog No. 5000239931
Encompass_Preferred
GSK8573 is a small-molecule research reagent used as a structurally related inactive control for acetyl-lysine competitive bromodomain inhibitors. It exhibits measurable binding to BRD9 (Kd ≈ 1.04 μM) and is supplied with characterization data and storage recommendations for reliable use in biochemical and cell-based assays.
  • High purity, supporting reproducible experimental results (reported ≈99.5%).
  • Validated as an inactive control for bromodomain inhibitor studies.
  • Measurable BRD9 binding, useful for target engagement comparisons.
  • Available in multiple pack sizes to suit assay throughput needs.
  • Detailed solubility and storage guidance for sample preparation and stability.
Catalog No. 50-002-39931 Supplier Medchemexpress LLC Supplier No. HY10747725MG
GSK8573 is a small-molecule research reagent used as a structurally related inactive control for acetyl-lysine competitive bromodomain inhibitors. It exhibits measurable binding to BRD9 (Kd ≈ 1.04 μM) and is supplied with characterization data and storage recommendations for reliable use in biochemical and cell-based assays.
  • High purity, supporting reproducible experimental results (reported ≈99.5%).
  • Validated as an inactive control for bromodomain inhibitor studies.
  • Measurable BRD9 binding, useful for target engagement comparisons.
  • Available in multiple pack sizes to suit assay throughput needs.
  • Detailed solubility and storage guidance for sample preparation and stability.