Learn More
Medchemexpress LLC 1-(1-(3-methoxyphenyl)-7-propoxyindolizin-3-yl)ethanone | 1693766-04-9 | MFCD30186082 | 99.5% | 323.39 | C20H21NO3 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10747750MG
Encompass_Preferred
GSK8573 is a small-molecule inactive control compound used in epigenetic reader-domain research and biochemical assays. Supplied as a solid for research use only, it is intended for use as a control in binding studies and compound screening. Store and handle according to the supplier safety data sheet and batch certificate of analysis.
Catalog No. 50-002-39939