Learn More
Medchemexpress LLC 1,1'-(4,4'-(((5-chloropyrimidine-2,4-diyl)bis(azanediyl))bis(3-methoxy-4,1-phenylene))bis(piperazine-4,1-diyl))diethanone | 1472795-20-2 | MFCD28133395 | 96.2% | 609.12 g·mol^-1 | C30H37ClN8O4 | 10MM 1ML
SDP
Catalog No. 5000239028
Encompass_Preferred
KRCA-0008 is a selective dual inhibitor of anaplastic lymphoma kinase (ALK) and activated Cdc42-associated kinase 1 (ACK1/TNK2) with reported IC50 values of approximately 12 nM for ALK and 4 nM for ACK1. It is intended for preclinical research into oncogenic signalling and cancer biology and is supplied as a concentrated DMSO solution suitable for in vitro biochemical and cellular assays.
  • Potent dual kinase inhibition with low nanomolar IC50 values.
  • Suitable for biochemical and cellular assays.
  • Provided as a 10 mM solution in DMSO for convenient dosing.
  • High purity (≈96.2%) with stated stability and storage recommendations.
  • Intended for research use only; not for human consumption.
Catalog No. 50-002-39028 Supplier Medchemexpress LLC Supplier No. HY1233110MM 1ML
KRCA-0008 is a selective dual inhibitor of anaplastic lymphoma kinase (ALK) and activated Cdc42-associated kinase 1 (ACK1/TNK2) with reported IC50 values of approximately 12 nM for ALK and 4 nM for ACK1. It is intended for preclinical research into oncogenic signalling and cancer biology and is supplied as a concentrated DMSO solution suitable for in vitro biochemical and cellular assays.
  • Potent dual kinase inhibition with low nanomolar IC50 values.
  • Suitable for biochemical and cellular assays.
  • Provided as a 10 mM solution in DMSO for convenient dosing.
  • High purity (≈96.2%) with stated stability and storage recommendations.
  • Intended for research use only; not for human consumption.