Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,4,4-Tetraethoxy-2-butyne 99%, Thermo Scientific™

Catalog No. AC241850010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC241850010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC241850010 Supplier Thermo Scientific Chemicals Supplier No. 241850010

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3-2-3988
Molecular Formula C13H8Br2O
Molecular Weight (g/mol) 340.01
MDL Number MFCD00016330
InChI Key LFABNOYDEODDFX-UHFFFAOYSA-N
Synonym 4,4'-dibromobenzophenone, bis 4-bromophenyl methanone, methanone, bis 4-bromophenyl, p,p'-dibromobenzophenone, bis p-bromophenyl ketone, 4,4-dibromobenzophenone, di4-bromophenyl ketone, bis-4-bromo-phenyl-methanone, zlchem 265, pubchem7494
PubChem CID 72868
IUPAC Name bis(4-bromophenyl)methanone
SMILES BrC1=CC=C(C=C1)C(=O)C1=CC=C(Br)C=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Yellow
Density 0.95
Infrared Spectrum Authentic
Refractive Index 1.4315 to 1.4335 (20°C, 589nm)
Linear Formula (C2H5O)2CHCidentical toCCH(OC2H5)2
Quantity 1 g
Formula Weight 230
Percent Purity ≥98.5%
Physical Form Liquid
Chemical Name or Material 1,1,4,4-Tetraethoxy-2-butyne, 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.