Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,4,4-Tetraphenyl-1,3-butadiene, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
10g
25g
Packaging:
Glass bottle
1450-63-1
C28H22
358.47
MFCD00004766
KLCLIOISYBHYDZ-UHFFFAOYSA-N
1,1,4,4-tetraphenyl-1,3-butadiene,tetraphenylbutadiene,1,1,4,4-tetraphenylbutadiene,1,1,4,4-tetraphenylbuta-1,3-diene,1,3-butadiene, 1,1,4,4-tetraphenyl,benzene, 1,1',1,1'-1,3-butadiene-1,4-diylidene tetrakis,tetraphenyl butadiene,1,1',1,1'-1,3-butadiene-1,4-diylidene tetrabenzene,1,4,4-triphenyl-1,3-butadienyl benzene,1,4,4-tetraphenylbutadiene
74060
1,4,4-triphenylbuta-1,3-dienylbenzene
C1=CC=C(C=C1)C(=CC=C(C2=CC=CC=C2)C3=CC=CC=C3)C4=CC=CC=C4
1450-63-1
C28H22
358.47
MFCD00004766
KLCLIOISYBHYDZ-UHFFFAOYSA-N
1,1,4,4-tetraphenyl-1,3-butadiene,tetraphenylbutadiene,1,1,4,4-tetraphenylbutadiene,1,1,4,4-tetraphenylbuta-1,3-diene,1,3-butadiene, 1,1,4,4-tetraphenyl,benzene, 1,1',1,1'-1,3-butadiene-1,4-diylidene tetrakis,tetraphenyl butadiene,1,1',1,1'-1,3-butadiene-1,4-diylidene tetrabenzene,1,4,4-triphenyl-1,3-butadienyl benzene,1,4,4-tetraphenylbutadiene
74060
1,4,4-triphenylbuta-1,3-dienylbenzene
C1=CC=C(C=C1)C(=CC=C(C2=CC=CC=C2)C3=CC=CC=C3)C4=CC=CC=C4
Packaging Glass bottle
Quantity 10g
Color White to Yellow
Melting Point 196.0°C to 200.0°C
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (HPLC)
Linear Formula (C6H5)2C=CHCH=C(C6H5)2
Beilstein 05, 750
Formula Weight 358.47
Physical Form Needle-Like Crystals
Percent Purity 99%
Chemical Name or Material 1,1,4,4-Tetraphenyl-1,3-butadiene, 99%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 215-914-7

RTECSNumber : CY9040630

TSCA : TSCA