Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,1,4,7,10,10-Hexamethyltriethylenetetramine

Catalog No. 36640425G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-640-425G 25 g
36-640-45G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 36-640-425G Supplier Sigma Aldrich Supplier No. 36640425G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1,1,4,7,10,10-hexamethyltriethylenetetramine is a polyamines additive, has been reported as an efficient reagent for the problematic koenigs-knorr glucuronidation.1,1,4,7,10,10-hexamethyltriethylenetetramine may be used as reagent in the synthesis of ideal linear random copolymers containing both vinyl polymer and polyester units in a single polymer chain
  • 1,1,4,7,10,10-hexamethyltriethylenetetramine complexed with cubr constitutes catalytic complex, used in the copolymerization of poly[α-caprolactone] with n,n-dimethylamino-2-ethyl methacrylate monomers by atom-transfer radical polymerization (atrp)
  • It may be used as catalyst in the aqueous surface-initiated-atrp to grow poly(n,n-dimethylacrylamide) (pdma)

Specifications

Density 0.847 g/mL (at 25°C (literature))
Boiling Point 130°C (11 mmHg)
Molecular Formula C12H30N4
Refractive Index n20/D 1.456 (literature)
Linear Formula [(CH3)2 NCH2CH2N(CH3)CH2-]2
MDL Number MFCD00025684
Quantity 25 g
Synonym HMTETA
Molecular Weight (g/mol) 230.39
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.