Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-[1-(4-Fluorobenzyl)-2-benzimidazolyl]-4-(methylamino)piperidine dihydrochloride, 97%, Thermo Scientific™

Catalog No. AAH63927MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63927MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH63927MD Supplier Thermo Scientific Chemicals Supplier No. H63927MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1134322-93-2
Molecular Formula C20H25Cl2FN4
Molecular Weight (g/mol) 411.346
MDL Number MFCD03844681
InChI Key ZIGFBTWAPHHQRA-UHFFFAOYSA-N
Synonym 1-1-4-fluorophenyl methyl-1h-benzimidazol-2-yl-n-methyl-4-piperidinamine dihydrochloride, 1-1-4-fluorophenyl methyl-1h-benzimidazol-2-yl-4-methylaminopiperidine dihydrochloride, 1-1-4-fluorophenyl methyl-1,3-benzodiazol-2-yl-n-methylpiperidin-4-amine dihydrochloride
PubChem CID 24720879
IUPAC Name 1-[1-[(4-fluorophenyl)methyl]benzimidazol-2-yl]-N-methylpiperidin-4-amine;dihydrochloride
SMILES CNC1CCN(CC1)C2=NC3=CC=CC=C3N2CC4=CC=C(C=C4)F.Cl.Cl

Specifications

Quantity 250 mg
Formula Weight 411.34
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-[1-(4-Fluorobenzyl)-2-benzimidazolyl]-4-(methylamino)piperidine dihydrochloride
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.