Learn More
Medchemexpress LLC 1-[1-(4-fluorophenyl)-4-(4-piperidinyl)butyl]pyrrolidin-2-one oxalate | 144734-36-1 | 98.9% | 429.48 g/mol | C24H28FNO5 | 5 MG
SDP
Catalog No. 5000211195
Encompass_Preferred
4F 4PP oxalate is the oxalate salt of 4-(4-fluorobenzoyl)-1-(4-phenylbutyl)piperidine, a selective 5-HT2A receptor antagonist with high affinity (Ki = 5.3 nM) and much lower affinity at 5-HT2C (Ki = 620 nM). Supplied as a solid and as ready-to-use DMSO solutions, it is intended for in vitro and in vivo receptor pharmacology research.
  • Selective 5-HT2A antagonist with high binding affinity.
  • Lower affinity at 5-HT2C reduces off-target activity.
  • High purity suitable for research applications.
  • Available as solid and as 10 mM DMSO solutions for convenience.
  • Soluble in DMSO with manufacturer-recommended in vivo formulations.
Catalog No. 50-002-11195 Supplier Medchemexpress LLC Supplier No. HY1009705MG
4F 4PP oxalate is the oxalate salt of 4-(4-fluorobenzoyl)-1-(4-phenylbutyl)piperidine, a selective 5-HT2A receptor antagonist with high affinity (Ki = 5.3 nM) and much lower affinity at 5-HT2C (Ki = 620 nM). Supplied as a solid and as ready-to-use DMSO solutions, it is intended for in vitro and in vivo receptor pharmacology research.
  • Selective 5-HT2A antagonist with high binding affinity.
  • Lower affinity at 5-HT2C reduces off-target activity.
  • High purity suitable for research applications.
  • Available as solid and as 10 mM DMSO solutions for convenience.
  • Soluble in DMSO with manufacturer-recommended in vivo formulations.