Learn More
Sigma Aldrich 1-(1-(4-Fluorophenyl)-5-methyl-1H-pyrazol-4-yl)ethanone
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- 3-ethoxythiophenol (etp) ) is a thiol derivative.3-ethoxythiophenol (etp) ) may be used to functionalize gold nanoparticles for developing:
- Partially selective gold nanoparticle chemiresistor sensors for the quantification of benzene, toluene, ethylbenzene, p-xylene and naphthalene (btexn) in water samples
- Flexible and tunable sensor based on monolayer-capped nanoparticles (mcnps), with potential application in e-skin devices
- Gold nanoparticle-based flexible sensor array
Specifications
Specifications
| Molecular Formula | C12H11FN2O |
| Linear Formula | C12H11FN2O |
| MDL Number | MFCD11168178 |
| Quantity | 1 g |
| Molecular Weight (g/mol) | 218.23 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.