Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-[1-(4-methoxyphenyl)-2-(methylamino)ethyl]cyclohexan-1-ol hydrochloride | 93413-90-2 | 99.0% | 299.84 g·mol⁻¹ | C16H25NO2·HCl | 5 MG
SDP

Catalog No. 5000262173
Encompass_Preferred

WY 45494 hydrochloride is the hydrochloride salt of N-desmethyl venlafaxine, a minor active metabolite and known impurity of the SNRI venlafaxine. It is supplied as a high-purity analytical standard for research, analytical method development, and impurity profiling.

  • High purity (99.0%).
  • Chemical formula C16H25NO2·HCl.
  • Molecular weight 299.84 g·mol⁻¹.
  • CAS number 93413-90-2.
  • Common pack sizes 5 mg and 25 mg.
  • Suitable as an analytical reference standard for impurity profiling and method development.
  • Useful for metabolism and synthetic chemistry studies.

Catalog No. 50-002-62173 Supplier Medchemexpress LLC Supplier No. HY131527R5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WY 45494 hydrochloride is the hydrochloride salt of N-desmethyl venlafaxine, a minor active metabolite and known impurity of the SNRI venlafaxine. It is supplied as a high-purity analytical standard for research, analytical method development, and impurity profiling.

  • High purity (99.0%).
  • Chemical formula C16H25NO2·HCl.
  • Molecular weight 299.84 g·mol⁻¹.
  • CAS number 93413-90-2.
  • Common pack sizes 5 mg and 25 mg.
  • Suitable as an analytical reference standard for impurity profiling and method development.
  • Useful for metabolism and synthetic chemistry studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.