Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-[1-(4-methoxyphenyl)-2-(methylamino)ethyl]cyclohexan-1-ol hydrochloride | 93413-90-2 | 99.6% | 299.83 g/mol | C16H26ClNO2 | 5 MG
SDP

Catalog No. 5000263991
Encompass_Preferred

High-purity hydrochloride salt of N-desmethyl venlafaxine used as a reference standard for analytical chemistry, impurity profiling, and method development in research laboratories.

  • Purity ~99.6%.
  • Molecular weight 299.83 g/mol.
  • Chemical formula C16H26ClNO2.
  • CAS number 93413-90-2.
  • Supplied in small-quantity vials suitable for reference standard use.
  • Intended for analytical and impurity identification applications.

Catalog No. 50-002-63991 Supplier Medchemexpress LLC Supplier No. HY1315275MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity hydrochloride salt of N-desmethyl venlafaxine used as a reference standard for analytical chemistry, impurity profiling, and method development in research laboratories.

  • Purity ~99.6%.
  • Molecular weight 299.83 g/mol.
  • Chemical formula C16H26ClNO2.
  • CAS number 93413-90-2.
  • Supplied in small-quantity vials suitable for reference standard use.
  • Intended for analytical and impurity identification applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.