Learn More
Medchemexpress LLC 1,1'-(9,9-dimethyl-9H-xanthene-4,5-diyl)bis(1,1-diphenylphosphine) | 161265-03-8 | MFCD00233866 | 99.9% | 578.62 g/mol | C39H32OP2 | 25g
SDP
Catalog No. 5000270143
Encompass_Preferred
Xantphos (9 9-Dimethyl-4 5-bis(diphenylphosphino)xanthene) is a bisphosphine ligand Xantphos coordinates to metals in either cis or trans chelation mode Xantphos acts as a key ligand in palladium or platinum catalyzed reactions Xantphos can be used in the construction of heterocyclic compounds in organic synthesis[1][2][3]
Catalog No. 50-002-70143 Supplier Medchemexpress LLC Supplier No. HYY009125G
Xantphos (9 9-Dimethyl-4 5-bis(diphenylphosphino)xanthene) is a bisphosphine ligand Xantphos coordinates to metals in either cis or trans chelation mode Xantphos acts as a key ligand in palladium or platinum catalyzed reactions Xantphos can be used in the construction of heterocyclic compounds in organic synthesis[1][2][3]