Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1'-Azobis(N,N-dimethylformamide), 98%

Catalog No. AC185710010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC185710010 1 g
AC185710050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC185710010 Supplier Thermo Scientific Chemicals Supplier No. 185710010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 10465-78-8 | C6H12N4O2 | 172.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10465-78-8
Molecular Formula C6H12N4O2
Molecular Weight (g/mol) 172.19
InChI Key VLSDXINSOMDCBK-BQYQJAHWSA-N
Synonym diamide, n,n,n',n'-tetramethylazodicarboxamide, tmad, 1,1'-azobis n,n-dimethylformamide, tetramethyldiazenedicarboxamide, diazenedicarboxamide, tetramethyl, n,n,n',n'-tetramethylazoformamide, diazenedicarboxylic acid bis n,n-dimethylamide, n,n,n',n'-tetramethylazobisformamide, tetramethylazodicarboxamide
PubChem CID 5353800
ChEBI CHEBI:48963
IUPAC Name (3E)-3-(dimethylcarbamoylimino)-1,1-dimethylurea
SMILES CN(C)C(=O)N=NC(=O)N(C)C

Specifications

Melting Point 111.0°C to 113.5°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Linear Formula (CH3)2NCON=NCON(CH3)2
Quantity 1 g
Beilstein 04,I,354
Formula Weight 172.19
Percent Purity 98%
Chemical Name or Material 1, 1'-Azobis(N, N-dimethylformamide)
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unw
EINECSNumber 233-951-7

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.