Learn More
Cayman Chemical 1,1'-(Azodicarbonyl)dipiperidine, Purity: ≥98%, CAS: 10465-81-3, 5 g
SDP
Catalog No. 502991141
Encompass_Preferred
A Mitsunobu reagent; has been used as a reagent in the synthesis of GPR120 agonists with antidiabetic activity and PPARα, PPARγ, and PPARδ triple agonists
Catalog No. 50-299-1141 Supplier Cayman Chemical Supplier No. 304865 G
A Mitsunobu reagent; has been used as a reagent in the synthesis of GPR120 agonists with antidiabetic activity and PPARα, PPARγ, and PPARδ triple agonists