Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(1-Benzofuran-3-yl)-2-bromo-1-ethanone, 95+%, Thermo Scientific™

Catalog No. CC27505DA
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC27505DA 1 g
CC27505CB 250 mg
CC27505DE 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC27505DA Supplier Thermo Scientific Chemicals Supplier No. CC27505DA
Add to Cart

Chemical Identifiers

CAS 187657-92-7
Molecular Formula C10H7BrO2
Molecular Weight (g/mol) 239.07
MDL Number MFCD03407369
InChI Key WVHFTONHSRLBGL-UHFFFAOYSA-N
Synonym 1-benzofuran-3-yl-2-bromoethanone, 1-1-benzofuran-3-yl-2-bromo-1-ethanone, 1-1-benzofuran-3-yl-2-bromoethanone, 1-benzofuran-3-yl-2-bromoethan-1-one, 1-1-benzofuran-3-yl-2-bromoethan-1-one, ethanone,1-3-benzofuranyl-2-bromo, 1-benzofuran-3-yl-2-bromo-1-ethanone, pubchem7014, 3-2-bromoacetyl benzo b furan, 1-benzofuran-3-yl-2-bromo-ethanone
PubChem CID 2776580
IUPAC Name 1-(1-benzofuran-3-yl)-2-bromoethanone
SMILES BrCC(=O)C1=COC2=CC=CC=C12

Specifications

Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 239.07
Percent Purity 95%
Chemical Name or Material 1-(1-Benzofuran-3-yl)-2-bromo-1-ethanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.