Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(1-Benzothiophen-5-yl)-2-bromo-1-ethanone, 97%, Thermo Scientific™

Catalog No. CC24805DE
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC24805DE 5 g Amber Glass Bottle
CC24805CB 250 mg Amber Glass Bottle
CC24805DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC24805DE Supplier Thermo Scientific Chemicals Supplier No. CC24805DE
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1131-87-9
Molecular Formula C10H7BrOS
Molecular Weight (g/mol) 255.13
MDL Number MFCD07368508
InChI Key NTQPGUCRHJHYIA-UHFFFAOYSA-N
Synonym 1-1-benzothiophen-5-yl-2-bromo-1-ethanone, 1-1-benzothiophen-5-yl-2-bromoethanone, 1-benzo b thiophen-5-yl-2-bromoethan-1-one, 5-bromoacetylbenzo b thiophene, 5-bromoacetyl-benzo b thiophene, 5-bromoacetyl benzo b thiophene, ethanone,1-benzo b thien-5-yl-2-bromo, 1-benzo b thiophen-5-yl-2-bromoethanone, 1-1-benzothiophen-5-yl-2-bromoethan-1-one
PubChem CID 7060546
IUPAC Name 1-(1-benzothiophen-5-yl)-2-bromoethanone
SMILES BrCC(=O)C1=CC=C2SC=CC2=C1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 255.13
Percent Purity 97%
Chemical Name or Material 1-(1-Benzothiophen-5-yl)-2-bromo-1-ethanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.