Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC [1,1'-Binaphthalene]-2,2'-diamine | 4488-22-6 | 284.36 | 25 G
SDP

Catalog No. 5000448236
Encompass_Preferred

1,1'-Binaphthyl-2,2'-diamine is an isomer of (S)-[1,1'-Binaphthalene]-2,2'-diamine, and can be utilized as an experimental control. (S)-[1,1'-Binaphthalene]-2,2'-diamine is a biochemical reagent suitable for use as a biological material or organic compound in life science-related research.

  • Off-white to light yellow solid
  • Molecular formula: C20H16N2
  • Molecular weight: 284.36
  • Purity (HPLC): 99.17%
  • Storage: 4°C, protected from light, stored under nitrogen
  • For research use only

Catalog No. 50-004-48236 Supplier Medchemexpress LLC Supplier No. 25GHYW007978B
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,1'-Binaphthyl-2,2'-diamine is an isomer of (S)-[1,1'-Binaphthalene]-2,2'-diamine, and can be utilized as an experimental control. (S)-[1,1'-Binaphthalene]-2,2'-diamine is a biochemical reagent suitable for use as a biological material or organic compound in life science-related research.

  • Off-white to light yellow solid
  • Molecular formula: C20H16N2
  • Molecular weight: 284.36
  • Purity (HPLC): 99.17%
  • Storage: 4°C, protected from light, stored under nitrogen
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.