Learn More
Medchemexpress LLC 1,1'-biphenyl-2,5-diol | 1079-21-6 | MFCD00002342 | 99.2% | 186.21 | C12H10O2 | 1 G
SDP
Catalog No. 5000302690
Encompass_Preferred
Phenylhydroquinone (2-phenylhydroquinone; CAS 1079-21-6) is a small-molecule research reagent that induces thymic atrophy and up-regulates apoptosis via p53 activation. It is supplied as a high-purity solid for use in in vitro and in vivo studies of apoptosis, thymic function, and related signaling pathways.
  • Orally active thymic atrophy inducer.
  • Modulates apoptosis through p53 activation.
  • High purity suitable for biological research.
  • Available in small-scale quantities for laboratory studies.
  • Chemically characterized: C12H10O2; MW 186.21.
Catalog No. 50-003-02690 Supplier Medchemexpress LLC Supplier No. HYW0161811G
Phenylhydroquinone (2-phenylhydroquinone; CAS 1079-21-6) is a small-molecule research reagent that induces thymic atrophy and up-regulates apoptosis via p53 activation. It is supplied as a high-purity solid for use in in vitro and in vivo studies of apoptosis, thymic function, and related signaling pathways.
  • Orally active thymic atrophy inducer.
  • Modulates apoptosis through p53 activation.
  • High purity suitable for biological research.
  • Available in small-scale quantities for laboratory studies.
  • Chemically characterized: C12H10O2; MW 186.21.