Learn More
Medchemexpress LLC 1,1'-biphenyl-2,5-diol | 1079-21-6 | MFCD00002342 | 99.2% | 186.21 g·mol⁻¹ | C12H10O2 | 10 G
SDP
Catalog No. 5000302290
Encompass_Preferred
Phenylhydroquinone (1,1'-biphenyl-2,5-diol) is a small-molecule research chemical used to induce thymic atrophy and promote p53-mediated apoptosis in preclinical studies. It is supplied as a solid with high purity for laboratory research into apoptosis and immune system modulation.
  • Orally active thymic atrophy inducer.
  • Up-regulates apoptosis through p53 activation.
  • High purity suitable for analytical and biological studies.
  • Solid form, off-white to gray, easy to handle and store.
  • Available in multiple lab-scale quantities for research use.
  • Provided with datasheet and certificate of analysis for batch-specific data.
Catalog No. 50-003-02290 Supplier Medchemexpress LLC Supplier No. HYW01618110G
Phenylhydroquinone (1,1'-biphenyl-2,5-diol) is a small-molecule research chemical used to induce thymic atrophy and promote p53-mediated apoptosis in preclinical studies. It is supplied as a solid with high purity for laboratory research into apoptosis and immune system modulation.
  • Orally active thymic atrophy inducer.
  • Up-regulates apoptosis through p53 activation.
  • High purity suitable for analytical and biological studies.
  • Solid form, off-white to gray, easy to handle and store.
  • Available in multiple lab-scale quantities for research use.
  • Provided with datasheet and certificate of analysis for batch-specific data.