Learn More
Medchemexpress LLC [1,1′-Biphenyl]-2,5-diol | 1079-21-6 | MFCD00002342 | 99.2% | 186.21 g/mol | C12H10O2 | 25 G
SDP
Catalog No. 5000303798
Encompass_Preferred
Phenylhydroquinone (PHQ) is a small-molecule research reagent reported to induce thymic atrophy and to up-regulate apoptosis through activation of p53. In cellular studies PHQ activates p53 via phosphorylation of Erk1/2 and p38 (example: 10 μM, 3 h in mouse thymocytes). It is supplied as a high-purity chemical for laboratory research.
  • Induces thymic atrophy and promotes apoptosis via p53 activation.
  • Activates p53 through phosphorylation of Erk1/2 and p38 in vitro.
  • High purity (99.21%) suitable for research applications.
  • Molecular weight 186.21 g/mol; formula C12H10O2.
  • Available in multiple laboratory pack sizes for experimental use.
  • Supplied with a certificate of analysis documenting purity.
Catalog No. 50-003-03798 Supplier Medchemexpress LLC Supplier No. 25GHYW016181
Phenylhydroquinone (PHQ) is a small-molecule research reagent reported to induce thymic atrophy and to up-regulate apoptosis through activation of p53. In cellular studies PHQ activates p53 via phosphorylation of Erk1/2 and p38 (example: 10 μM, 3 h in mouse thymocytes). It is supplied as a high-purity chemical for laboratory research.
  • Induces thymic atrophy and promotes apoptosis via p53 activation.
  • Activates p53 through phosphorylation of Erk1/2 and p38 in vitro.
  • High purity (99.21%) suitable for research applications.
  • Molecular weight 186.21 g/mol; formula C12H10O2.
  • Available in multiple laboratory pack sizes for experimental use.
  • Supplied with a certificate of analysis documenting purity.