Learn More
Medchemexpress LLC [1,1′-Biphenyl]-2,5-diol | 1079-21-6 | MFCD00002342 | 99.2% | 186.21 g·mol⁻¹ | C12H10O2 | 50 G
SDP
Catalog No. 5000302176
Encompass_Preferred
Phenylhydroquinone (2-Phenylhydroquinone) is a small-molecule research reagent reported to induce thymic atrophy and to up-regulate apoptosis via p53 activation. It is used in studies of apoptosis-related signaling and thymic atrophy mechanisms, with reported activity involving MDM2/p53, ERK, and p38 MAPK pathways.
  • Orally active thymic atrophy inducer suitable for in vivo and in vitro research.
  • Up-regulates apoptosis through activation of p53.
  • Engages MDM2/p53, ERK, and p38 MAPK pathways relevant to apoptosis studies.
  • High purity (99.21%) appropriate for experimental applications.
  • Chemical formula C12H10O2; molecular weight 186.21 g·mol⁻¹; CAS 1079-21-6.
Catalog No. 50-003-02176 Supplier Medchemexpress LLC Supplier No. HYW01618150G
Phenylhydroquinone (2-Phenylhydroquinone) is a small-molecule research reagent reported to induce thymic atrophy and to up-regulate apoptosis via p53 activation. It is used in studies of apoptosis-related signaling and thymic atrophy mechanisms, with reported activity involving MDM2/p53, ERK, and p38 MAPK pathways.
  • Orally active thymic atrophy inducer suitable for in vivo and in vitro research.
  • Up-regulates apoptosis through activation of p53.
  • Engages MDM2/p53, ERK, and p38 MAPK pathways relevant to apoptosis studies.
  • High purity (99.21%) appropriate for experimental applications.
  • Chemical formula C12H10O2; molecular weight 186.21 g·mol⁻¹; CAS 1079-21-6.