Learn More
Medchemexpress LLC [1,1'-Biphenyl]-3,3',4,4'-tetrol | 3598-30-9 | MFCD00038783 | 98.3% | 218.21 | C12H10O4 | 1 ML
SDP
Catalog No. 5000285265
Encompass_Preferred
Biphenyl-3′,3,4,4′-tetrol (BPT) is a small-molecule research compound reported to inhibit Aβ40 aggregation and used in studies of neurodegenerative diseases such as Alzheimer's disease. Available as a solid or as a 10 mM solution in DMSO (1 mL).
  • Inhibits Aβ40 aggregation, used for neurodegeneration research.
  • Available as a 10 mM solution in DMSO (1 mL) or as a solid for synthesis and assays.
  • Molecular formula C12H10O4, molecular weight 218.21 g/mol.
  • High purity (≈98.3% by HPLC) suitable for research applications.
  • Off-white to gray solid appearance.
  • Store under nitrogen; in solvent: -80°C (6 months) or -20°C (1 month).
Catalog No. 50-002-85265 Supplier Medchemexpress LLC Supplier No. HYW59485110MM/1ML
Biphenyl-3′,3,4,4′-tetrol (BPT) is a small-molecule research compound reported to inhibit Aβ40 aggregation and used in studies of neurodegenerative diseases such as Alzheimer's disease. Available as a solid or as a 10 mM solution in DMSO (1 mL).
  • Inhibits Aβ40 aggregation, used for neurodegeneration research.
  • Available as a 10 mM solution in DMSO (1 mL) or as a solid for synthesis and assays.
  • Molecular formula C12H10O4, molecular weight 218.21 g/mol.
  • High purity (≈98.3% by HPLC) suitable for research applications.
  • Off-white to gray solid appearance.
  • Store under nitrogen; in solvent: -80°C (6 months) or -20°C (1 month).