Learn More
Medchemexpress LLC [1,1'-Biphenyl]-3-carboxamide, 5-cyano-2'-[4-[2-(3-methyl-1H-indol-1-yl)ethyl]-1-piperazinyl]- | 1793053-37-8 | 99.1% | 574.76 | 1 ML
SDP
Catalog No. 5000385825
Encompass_Preferred
LLY-507 is a potent and selective inhibitor of protein-lysine methyltransferase SMYD2. It potently inhibits the ability of SMYD2 to methylate p53 peptide with an IC50 <15 nM. It serves as a valuable chemical probe to aid in the dissection of SMYD2 function in cancer and other biological processes.
  • Potent and selective inhibitor of protein-lysine methyltransferase SMYD2.
  • Inhibits SMYD2 to methylate p53 peptide with an IC50 <15 nM.
  • Valuable chemical probe for dissecting SMYD2 function in cancer and other biological processes.
Catalog No. 50-003-85825 Supplier Medchemexpress LLC Supplier No. HY1931310MM/1ML
LLY-507 is a potent and selective inhibitor of protein-lysine methyltransferase SMYD2. It potently inhibits the ability of SMYD2 to methylate p53 peptide with an IC50 <15 nM. It serves as a valuable chemical probe to aid in the dissection of SMYD2 function in cancer and other biological processes.
  • Potent and selective inhibitor of protein-lysine methyltransferase SMYD2.
  • Inhibits SMYD2 to methylate p53 peptide with an IC50 <15 nM.
  • Valuable chemical probe for dissecting SMYD2 function in cancer and other biological processes.