Learn More
Medchemexpress LLC [1,1'-Biphenyl]-3-propanoic acid, a-amino-2'-chloro-5-(phosphonomethyl)-, (aS)- | 174575-17-8 | 98.8% | 1 ML
SDP
Catalog No. 5000338180
Encompass_Preferred
SDZ 220-581 is an orally active, potent, and competitive NMDA receptor antagonist with a pKi value of 7.7. In vivo studies have demonstrated that it dose-dependently protects against maximal electroshock seizures (MES), exhibiting a rapid onset and long duration of action.
  • Orally active NMDA receptor antagonist
  • Potent and competitive action
  • pKi value of 7.7
  • Protects against maximal electroshock seizures
  • Rapid onset and long duration of action
  • For research use only
Catalog No. 50-003-38180 Supplier Medchemexpress LLC Supplier No. HY1305910MM/1ML
SDZ 220-581 is an orally active, potent, and competitive NMDA receptor antagonist with a pKi value of 7.7. In vivo studies have demonstrated that it dose-dependently protects against maximal electroshock seizures (MES), exhibiting a rapid onset and long duration of action.
  • Orally active NMDA receptor antagonist
  • Potent and competitive action
  • pKi value of 7.7
  • Protects against maximal electroshock seizures
  • Rapid onset and long duration of action
  • For research use only