Learn More
Medchemexpress LLC [1,1'-Biphenyl]-3-propanoic acid, a-amino-2'-chloro-5-(phosphonomethyl)-, (aS)- | 174575-17-8 | 98.8% | 25 MG
SDP
Catalog No. 5000341496
Encompass_Preferred
SDZ 220-581 is an orally active, potent, competitive NMDA receptor antagonist with a pKi value of 7.7. It has been shown to dose-dependently protect mice against maximal electroshock seizures (MES) upon oral administration, with a rapid onset and long duration of action.
  • Potent, competitive NMDA receptor antagonist
  • Orally active
  • Demonstrated pKi value of 7.7
  • Protects against maximal electroshock seizures (MES) in vivo
  • Offers rapid onset of action
  • Provides long duration of action
  • High purity of 98.76%
Catalog No. 50-003-41496 Supplier Medchemexpress LLC Supplier No. 25MGHY13059
SDZ 220-581 is an orally active, potent, competitive NMDA receptor antagonist with a pKi value of 7.7. It has been shown to dose-dependently protect mice against maximal electroshock seizures (MES) upon oral administration, with a rapid onset and long duration of action.
  • Potent, competitive NMDA receptor antagonist
  • Orally active
  • Demonstrated pKi value of 7.7
  • Protects against maximal electroshock seizures (MES) in vivo
  • Offers rapid onset of action
  • Provides long duration of action
  • High purity of 98.76%