Learn More
Medchemexpress LLC [1,1'-Biphenyl]-3-propanoic acid, α-amino-2'-chloro-5-(phosphonomethyl)-, (αS)- | 174575-17-8 | 98.8% | 369.74 g/mol | C16H17ClNO5P | 5 MG
SDP
Catalog No. 5000262217
Encompass_Preferred
SDZ 220-581 is an orally active, potent, competitive NMDA receptor antagonist (pKi = 7.7) used as a research compound to probe NMDA receptor pharmacology and anticonvulsant activity. It has demonstrated dose-dependent protection against maximal electroshock seizures in mice and is supplied as a light yellow to yellow solid.
  • Potent competitive NMDA receptor antagonist with reported pKi = 7.7.
  • Orally active and effective in in vivo seizure models.
  • Suitable for in vitro receptor pharmacology and in vivo efficacy studies.
  • Provided as a light yellow solid; available in small milligram pack sizes.
  • Molecular weight 369.74 g/mol and formula C16H17ClNO5P.
Catalog No. 50-002-62217 Supplier Medchemexpress LLC Supplier No. HY130595MG
SDZ 220-581 is an orally active, potent, competitive NMDA receptor antagonist (pKi = 7.7) used as a research compound to probe NMDA receptor pharmacology and anticonvulsant activity. It has demonstrated dose-dependent protection against maximal electroshock seizures in mice and is supplied as a light yellow to yellow solid.
  • Potent competitive NMDA receptor antagonist with reported pKi = 7.7.
  • Orally active and effective in in vivo seizure models.
  • Suitable for in vitro receptor pharmacology and in vivo efficacy studies.
  • Provided as a light yellow solid; available in small milligram pack sizes.
  • Molecular weight 369.74 g/mol and formula C16H17ClNO5P.