Learn More
Medchemexpress LLC 1,1'-Biphenyl, 4-methyl- | 644-08-6 | MFCD00008544 | 100.0% | 168.23 g/mol | C13H12 | 100 G
SDP
Catalog No. 5000299486
Encompass_Preferred
4-Methylbiphenyl (1,1'-biphenyl, 4-methyl-) is an aromatic hydrocarbon and endogenous metabolite used as a reference standard and reagent in analytical and biochemical research. Supplied as a white to off-white solid with high purity, it is suitable for analytical applications and formulation in organic solvents.
  • High purity for analytical use (99.98%).
  • White to off-white solid form.
  • Soluble in common organic solvents; DMSO recommended.
  • Available in multiple pack sizes, including 100 g.
  • COA and SDS available for quality and safety verification.
Catalog No. 50-002-99486 Supplier Medchemexpress LLC Supplier No. HYW017077100G
4-Methylbiphenyl (1,1'-biphenyl, 4-methyl-) is an aromatic hydrocarbon and endogenous metabolite used as a reference standard and reagent in analytical and biochemical research. Supplied as a white to off-white solid with high purity, it is suitable for analytical applications and formulation in organic solvents.
  • High purity for analytical use (99.98%).
  • White to off-white solid form.
  • Soluble in common organic solvents; DMSO recommended.
  • Available in multiple pack sizes, including 100 g.
  • COA and SDS available for quality and safety verification.