Learn More
Medchemexpress LLC [1,1'-Biphenyl]-4-pentanoic acid, γ-[(3-carboxy-1-oxopropyl)amino]-α-methyl-, (αR,γS)-d4 | 149709-44-4 | 99.3% | 387.46 | 5 MG
SDP
Catalog No. 5000386966
Encompass_Preferred
Sacubitrilat-d4 is the deuterium-labeled Sacubitrilat, an active neprilysin (NEP) inhibitor. Deuteration, the incorporation of stable heavy isotopes into drug molecules, is used for quantitation during drug development and can affect pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-86966 Supplier Medchemexpress LLC Supplier No. HY17620S5MG
Sacubitrilat-d4 is the deuterium-labeled Sacubitrilat, an active neprilysin (NEP) inhibitor. Deuteration, the incorporation of stable heavy isotopes into drug molecules, is used for quantitation during drug development and can affect pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS