Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC [1,1'-Biphenyl]-4-ylboronic acid | 5122-94-1 | 198.03 | 100 G
SDP

Catalog No. 5000445678
Encompass_Preferred

[1,1'-Biphenyl]-4-ylboronic acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is for research use only.

  • Appearance: Solid
  • Color: White to off-white
  • Solubility (In Vitro): DMSO: 100 mg/mL (504.97 mM; requires ultrasonic; hygroscopic DMSO has a significant impact on solubility, use newly opened DMSO)
  • Antagonist activity at recombinant human TRPV1 channel expressed in HEK293 cells assessed as inhibition of capsaicin-induced Ca2+ flux, preincubated for 5 minutes followed by capsaicin challenge by Fluo-4-AM dye based spectrofluorimetry.

Catalog No. 50-004-45678 Supplier Medchemexpress LLC Supplier No. HYW002364100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

[1,1'-Biphenyl]-4-ylboronic acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is for research use only.

  • Appearance: Solid
  • Color: White to off-white
  • Solubility (In Vitro): DMSO: 100 mg/mL (504.97 mM; requires ultrasonic; hygroscopic DMSO has a significant impact on solubility, use newly opened DMSO)
  • Antagonist activity at recombinant human TRPV1 channel expressed in HEK293 cells assessed as inhibition of capsaicin-induced Ca2+ flux, preincubated for 5 minutes followed by capsaicin challenge by Fluo-4-AM dye based spectrofluorimetry.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.