Learn More
Medchemexpress LLC [1,1'-Biphenyl]-4-ylboronic acid | 5122-94-1 | 198.03 | 50 G
SDP
Catalog No. 5000445885
Encompass_Preferred
[1,1'-Biphenyl]-4-ylboronic acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is a boric acid compound, appearing as a white to off-white solid.
  • Biochemical reagent for life science related research
  • Can be used as a biological material or organic compound
  • Appears as a solid, white to off-white in color
  • Exhibits antagonist activity at recombinant human TRPV1 channels in HEK293 cells with an IC50 > 10 μM
  • For research use only
Catalog No. 50-004-45885 Supplier Medchemexpress LLC Supplier No. HYW00236450G
[1,1'-Biphenyl]-4-ylboronic acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is a boric acid compound, appearing as a white to off-white solid.
  • Biochemical reagent for life science related research
  • Can be used as a biological material or organic compound
  • Appears as a solid, white to off-white in color
  • Exhibits antagonist activity at recombinant human TRPV1 channels in HEK293 cells with an IC50 > 10 μM
  • For research use only