Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1'-Bis(dichlorophosphino)ferrocene 95.0+%, TCI America™
SDP

Catalog No. B4272200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4272200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B4272200MG Supplier TCI America Supplier No. B4272200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 142691-70-1
Melting Point 70°C
Molecular Formula C10H8Cl4FeP2
Quantity 200 mg
MDL Number MFCD01630854
UN Number 3261
SMILES [Fe].ClP(Cl)c1cccc1.ClP(Cl)c1cccc1
IUPAC Name 1,1'-Bis(dichlorophosphino)ferrocene
Molecular Weight (g/mol) 387.77
Formula Weight 387.77
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,1′-Bis(dichlorophosphino)ferrocene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.