Learn More
Strem, An Ascensus Company CAS# 146960-90-9. 1,1'-Bis(dicyclohexylphosphino)ferrocene, min. 98% MFCD09038820. 2G
SDP
Supplier:  Strem, An Ascensus Company 2601552G
Encompass
CAS# 146960-90-9. 2G. 1,1'-Bis(dicyclohexylphosphino)ferrocene, min. 98%. MFCD09038820. Molecular Formula: C34H52FeP2 Molecular Weight: 578.57. Color/Form: orange pwdr. Stability: air sensitive. Strem#: 26-0155 www.strem.com/catalog/v3/26-0155/
Catalog No. 50-138-5452