Learn More
Strem, An Ascensus Company CAS# 12150-46-8. 1g. 1,1'-Bis(diphenylphosphino)ferrocene, 99% DPPF. MFCD00001422
SDP
Supplier:  Strem, An Ascensus Company 2602701G
Encompass
CAS# 12150-46-8. 1g. 1,1'-Bis(diphenylphosphino)ferrocene, 99% DPPF. MFCD00001422. Molecular Weight: 554.39. Molecular Formula: (C6H5)2PC5H4FeC5H4P(C6H5)2. Color/form: yellow to orange xtl. Strem# 26-0270. http://www.strem.com/catalog/v/26-0270/
Catalog No. 50-901-10970