Learn More
AdipoGen 1,1-Bis(phenylsulfonyl)ethylene (1 g)
SDP
Supplier:  AdipoGen CDXB0127G001
Encompass_Preferred
1,1-Bis(phenylsulfonyl)ethylene (1 g), AdipoGen Life Sciences. Chemical. CAS: 39082-53-6. Formula: C14H12O4S2. MW: 308.4. Synthetic. Synthetic compound equivalent to the ethylene 1,2-dipole. Dienophile used for Diels-Alder reactions and useful synthon for neutral homologation of ketones. Used in the preparation of alpha,alpha-disubstituted alpha-amino acid derivatives.
Catalog No. 50-149-0256