Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1-Bis(trimethylsilyloxy)-ethene, Thermo Scientific™

Catalog No. AC331000010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC331000010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC331000010 Supplier Thermo Scientific Chemicals Supplier No. 331000010

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24697-35-6
Molecular Formula C8H20O2Si2
Molecular Weight (g/mol) 204.416
MDL Number MFCD01632562
InChI Key QTLOQPVMBORRRD-UHFFFAOYSA-N
Synonym ketene bis trimethylsilyl acetal, 2,2,6,6-tetramethyl-4-methylidene-3,5-dioxa-2,6-disilaheptane, 1,1-bis trimethylsilyloxy ethene, 1,1-bis trimethylsilyloxy-ethene, trimethyl 1-trimethylsilyloxyethenoxy silane
PubChem CID 2733564
IUPAC Name trimethyl(1-trimethylsilyloxyethenoxy)silane
SMILES C[Si](C)(C)OC(=C)O[Si](C)(C)C

Specifications

Color Colorless to Yellow
Infrared Spectrum Authentic
Refractive Index 1.4180 to 1.4210 (20°C, 589nm)
Linear Formula CHC(OSi(CH3)3)
Quantity 1 g
Formula Weight 204.42
Physical Form Liquid
Chemical Name or Material 1, 1-Bis(trimethylsilyloxy)-ethene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.