Learn More
AdipoGen 1 1-Bisphenylsulfonylethylen
SDP
Supplier:  AdipoGen CDXB0127G005
Encompass_Preferred
Chemical. CAS 39082-53-6. Formula C14H12O4S2. MW 308.4. Synthetic. Synthetic compound equivalent to the ethylene 1,2-dipole. Dienophile used for Diels-Alder reactions and useful synthon for neutral homologation of ketones. Used in the preparation of alpha,alpha-disubstituted alpha-amino acid derivatives.
Catalog No. 50-256-1663