Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(1-Boc-4-piperidinyl)-4-iodopyrazole, tech. 90%, Thermo Scientific™

Catalog No. AAH6591806
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6591806 5 g
AAH65918MD 250 mg
AAH6591803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6591806 Supplier Thermo Scientific Chemicals Supplier No. H6591806

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 877399-73-0
Molecular Formula C13H20IN3O2
Molecular Weight (g/mol) 377.226
MDL Number MFCD12913927
InChI Key IONGFFYQZADMPH-UHFFFAOYSA-N
Synonym tert-butyl 4-4-iodo-1h-pyrazol-1-yl piperidine-1-carboxylate, 1-boc-4-4-iodo-1h-pyrazol-1-yl piperidine, tert-butyl 4-4-iodopyrazol-1-yl piperidine-1-carboxylate, 1-1-boc-4-piperidinyl-4-iodopyrazole, tert-butyl4-4-iodo-1h-pyrazol-1-yl piperidine-1-carboxylate, 1-piperidinecarboxylic acid, 4-4-iodo-1h-pyrazol-1-yl-, 1,1-dimethylethyl ester, 1-n-boc-4-4-iodopyrazol-1-yl piperidine, 1-4-boc-piperidino-4-iodopyrazole, 1-n-boc-4-4-iodo-1h-pyrazol-1-yl piperidine, 1-1-tert-butoxycarbonyl-4-piperidinyl-4-iodopyrazole
PubChem CID 45480389
IUPAC Name tert-butyl 4-(4-iodopyrazol-1-yl)piperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(CC1)N2C=C(C=N2)I

Specifications

Quantity 5 g
Beilstein 14511019
Sensitivity Light sensitive
Formula Weight 377.23
Percent Purity 90%
Grade Technical
Physical Form Solid
Chemical Name or Material 1-(1-Boc-4-piperidinyl)-4-iodopyrazole
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.