Learn More
Medchemexpress LLC 1,1'-(Butane-1,4-diyl)bis(3-(2-(2-(2-aminoethoxy)ethoxy)ethyl)urea) | 1571103-88-2 | 99.2% | 509.47 | 1 G
SDP
Catalog No. 5000404387
Encompass_Preferred
1,1'-(Butane-1,4-diyl)bis(3-(2-(2-(2-aminoethoxy)ethoxy)ethyl)urea) dihydrochloride is a drug intermediate for the synthesis of various active compounds. It is intended for research use only.
  • Appears as a solid.
  • Ships at room temperature in continental US.
  • Store at 4°C, sealed, away from moisture.
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month, sealed and away from moisture.
Catalog No. 50-004-04387 Supplier Medchemexpress LLC Supplier No. HY424971G
1,1'-(Butane-1,4-diyl)bis(3-(2-(2-(2-aminoethoxy)ethoxy)ethyl)urea) dihydrochloride is a drug intermediate for the synthesis of various active compounds. It is intended for research use only.
  • Appears as a solid.
  • Ships at room temperature in continental US.
  • Store at 4°C, sealed, away from moisture.
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month, sealed and away from moisture.