Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1'-Carbonyldiimidazole 97.0+%, TCI America™
SDP

Catalog No. C011925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C011925G 25 g
C01195G 5 g
C0119250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C011925G Supplier TCI America Supplier No. C011925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 530-62-1
Molecular Formula C7H6N4O
Molecular Weight (g/mol) 162.15
MDL Number MFCD00005286
InChI Key PFKFTWBEEFSNDU-UHFFFAOYSA-N
Synonym 1,1'-carbonyldiimidazole, n,n'-carbonyldiimidazole, carbonyldiimidazole, di 1h-imidazol-1-yl methanone, n,n-carbonyldiimidazole, 1h-imidazole, 1,1'-carbonylbis, 1,1'-carbonylbis-1h-imidazole, carbonyl diimidazole, diimidazol-1-yl ketone, 1,1-carbonyldiimidazole
PubChem CID 68263
IUPAC Name 1-(1H-imidazole-1-carbonyl)-1H-imidazole
SMILES O=C(N1C=CN=C1)N1C=CN=C1

Specifications

Melting Point 118°C
Color White-Yellow
pH 8
Quantity 25 g
UN Number 1759
Formula Weight 162.15
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,1′-Carbonyldiimidazole [Coupling Agent for Peptides Synthesis]
EINECSNumber (5)-6491
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.