Learn More
AdipoGen 1 1-Carbonyldipyrrolidine
SDP
Supplier:  AdipoGen CDXC0780G050
Encompass_Preferred
Chemical. CAS 81759-25-3. Formula C9H16N2O. MW 168.24. Used as building block and intermediate in organic synthesis and as coupling reagent in peptide synthesis.
Catalog No. 50-256-1335